Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TLB 150 Benzoate | 1208070-53-4 | 98.3% | C27H20ClN5O3 | 1 MG
SDP

Catalog No. 5000359073
Encompass_Preferred

TLB 150 Benzoate (RAD150) is a modulator for androgen receptor with an IC50 of 0.13 μM. It is intended for research use only and not for patient administration.

  • Modulator for androgen receptor
  • IC50 of 0.13 μM
  • Molecular weight: 497.93
  • Appearance: White to off-white solid
  • Storage: Powder at -20°C for 3 years, in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-59073 Supplier Medchemexpress LLC Supplier No. HY455091MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLB 150 Benzoate (RAD150) is a modulator for androgen receptor with an IC50 of 0.13 μM. It is intended for research use only and not for patient administration.

  • Modulator for androgen receptor
  • IC50 of 0.13 μM
  • Molecular weight: 497.93
  • Appearance: White to off-white solid
  • Storage: Powder at -20°C for 3 years, in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.