Learn More
Medchemexpress LLC TLB 150 Benzoate (RAD150) | 1208070-53-4 | 98.3% | C27H20ClN5O3 | 5 MG
SDP
Catalog No. 5000359075
Encompass_Preferred
TLB 150 Benzoate (RAD150) is a modulator for the androgen receptor with an IC50 of 0.13 μM.
  • AR modulator
  • Modulator for androgen receptor with an IC50 of 0.13 μM
Catalog No. 50-003-59075 Supplier Medchemexpress LLC Supplier No. HY455095MG
TLB 150 Benzoate (RAD150) is a modulator for the androgen receptor with an IC50 of 0.13 μM.
  • AR modulator
  • Modulator for androgen receptor with an IC50 of 0.13 μM