Learn More
Miles Scientific RP18F ON GLASS 250UM 20X20CM (25 PLATES) 
SDP
Catalog No. NC0059065
Encompass_Preferred
Reversed Phase Octadecyl Silanized Silica Gel. These reversed phase TLC plates have been manufacturer to be greater than 50% and less than 100% silanized. Because of this they are hydrophobic, but wettable (unlike CAT# 164077 & 165077 which are 100% silanized). FEATURES: 14% carbon content - Acid stable (UV254) fluorescent indicator - 20x20 cm glass backing - 250um coating thickness - 25 plates/box - Recomended Applications: aminophenols, barbiturates, preservatives, nucleobases, polycyclic aromatic hydrocarbons, steroids, tetracyclines, plasticizers (phthalates)
Catalog No. NC0059065 Supplier Miles Scientific Supplier No. P173011
Reversed Phase Octadecyl Silanized Silica Gel. These reversed phase TLC plates have been manufacturer to be greater than 50% and less than 100% silanized. Because of this they are hydrophobic, but wettable (unlike CAT# 164077 & 165077 which are 100% silanized). FEATURES: 14% carbon content - Acid stable (UV254) fluorescent indicator - 20x20 cm glass backing - 250um coating thickness - 25 plates/box - Recomended Applications: aminophenols, barbiturates, preservatives, nucleobases, polycyclic aromatic hydrocarbons, steroids, tetracyclines, plasticizers (phthalates)