Learn More
BRINSEA PRODUCTS BRINSEA PRODUCTS TLC-50 Advance Series II Bird Brooder/Intensive Care Unit/Recovery Incubator
Supplier:  BRINSEA PRODUCTS USHD4111C
Encompass_Preferred
Tough, efficient, easily cleaned and quick to set up, the Brinsea TLC-50 Advance Series II Bird Brooder/Animal Recovery Incubator is a multi-purpose Intensive Care Unit providing optimal brooding of baby parrots and other newly hatched exotic chicks and ideal recovery conditions for sick, injured or orphaned birds and mammals. It comes with many sophisticated features which include: Accurate digital temperature control adjustable through an easy to use menu with display in °C or °F; High and low temperature alarms including Brinsea’s patented room temperature alarm; Digital display of the humidity level achieved in the chamber; Water reservoir with external top up to reduce the risk of dehydration; Unique air filtration for removal of airborne bacteria and fungi from the chamber (filter replacements available); Adjustable fresh air exchange through a door mounted vent; Variable fan speed for particularly stress-sensitive animals;
Catalog No. NC2224713