Learn More
Atmar Glass Company TLC CHAMBER
Catalog No. NC9774232
Encompass
Tlc Chamber With Vinyl Cap 59mm Id X 125mm High
Catalog No. NC9774232 Supplier Atmar Glass Company Supplier No. A1001621
Tlc Chamber With Vinyl Cap 59mm Id X 125mm High