Learn More
Chemglass Life Sciences TLC Developing Chamber Complete, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 8
SDP
Catalog No. CG118104
Encompass_Preferred
TLC Developing Chamber, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 8. Chamber has a short length standard taper outer joint. Stopper is a short length inner with a tool ball style handle. Complete chamber is supplied with bottle and stopper.
Catalog No. CG118104 Supplier Chemglass Life Sciences Supplier No. CG118104
TLC Developing Chamber, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 8. Chamber has a short length standard taper outer joint. Stopper is a short length inner with a tool ball style handle. Complete chamber is supplied with bottle and stopper.