Learn More
Chemglass Life Sciences TLC Developing Chamber Complete, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 4
SDP
Catalog No. CG118103
Encompass_Preferred
TLC Developing Chamber, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 4. Chamber has a short length standard taper outer joint. Stopper is a short length inner with a tool ball style handle. Complete chamber is supplied with bottle and stopper.
Catalog No. CG118103 Supplier Chemglass Life Sciences Supplier No. CG118103
TLC Developing Chamber, Cylindrical, 60/15 Joint Size, Fits Plate 2in X 4. Chamber has a short length standard taper outer joint. Stopper is a short length inner with a tool ball style handle. Complete chamber is supplied with bottle and stopper.