Learn More
MilliporeSigma™ TLC and HPTLC GLP Plates, Glass Backed
Catalog No. M1055640001 Shop All MilliporeSigma Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
10 x 20 cm
20 x 20 cm
Type:
GLP Silica TLC Plate
HPTLC GLP Silica Gel 60 F254
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Type
M1055640001 10 x 10 cm HPTLC GLP Silica Gel 60 F254
M1055660001 20 x 20 cm GLP Silica TLC Plate
M1057020001 10 x 20 cm HPTLC GLP Silica Gel 60 F254
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1055640001 Supplier MilliporeSigma™ Supplier No. 1055640001
Add to Cart
Add to Cart
For thin layer chromatography applications.
These plates feature individual laser coding for GLP applications. Designed for GLP applications, the tops of each of these plates bear an item, batch, and individual plate number. As a result, lab professionals can easily record and archive every plate they use. Based on our proven silica found in TLC and HPTLC plates, these GLP plates deliver equally unsurpassed separation performance.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 Plates
Type HPTLC GLP Silica Gel 60 F254
For Use With (Application) GLP Applications
Dimensions (L x W) 10 x 10 cm