Learn More
MilliporeSigma™ Supelco™ TLC Plate Storage Racks
Catalog No. 111009367 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Holds:
20 x 20 cm Plates
Catalog No. Holds
11-100-9367 20 x 20 cm Plates
1 options
Catalog No. 11-100-9367 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2660941EA
Add to Cart
Add to Cart
Used to carry and store TLC plates without damage to media.
Racked plates may be air or oven dried. Holds up to 10 standard plates in either a vertical or horizontal position. Racks are stainless steel with a carrying handle.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (20 Gauge)
For Use With (Application) TLC
Holds 20 x 20 cm Plates