Learn More
Phenomenex Inc TLC Plates
Catalog No. 502412670 Shop All Phenomenex Products
Encompass_Preferred
Silica; 200x200mm; 60; 10ea/pk; Glass; S; F254; THK 1.0mm
Catalog No. 50-241-2670 Supplier Phenomenex Inc Supplier No. TZCSF2002000
Silica; 200x200mm; 60; 10ea/pk; Glass; S; F254; THK 1.0mm