Learn More
Phenomenex Inc TLC Plates
Catalog No. 502412856 Shop All Phenomenex Products
Encompass_Preferred
Silica; 200x200mm; 60; 10ea/pk; Glass; M; F254
Catalog No. 50-241-2856 Supplier Phenomenex Inc Supplier No. TCSF200200M
Silica; 200x200mm; 60; 10ea/pk; Glass; M; F254