Learn More
Miles Scientific SILICA GEL G 2000UM 20X20CM (25 PLATES/BOX) 
SDP
Catalog No. NC1120880
Encompass_Preferred
Preparative TLC plates have adsorbent layers thicker than the 250 micron layers normally used for analytical separations. Thus, preparative plates can be used for separation of larger quantities of materials. The loading capacity for a given degree of sample component resolution increases roughly as the square root of the increased adsorbent layer thickness. In preparative TLC, materials to be separated are often applied as long streaks rather than spots. After development, specific compounds may be recovered by scraping the adsorbent layers from the plate in the region of interest and eluting the separated material from the adsorbent using a strong solvent. Thicker layers for higher capacity -Full range of plate sizes ·Prep-scored option for convenient zone location - Available with preadsorbent sample loading zone - Calcium sulfate binder-soft later for easy scraping
Catalog No. NC1120880 Supplier Miles Scientific Supplier No. P01015
Preparative TLC plates have adsorbent layers thicker than the 250 micron layers normally used for analytical separations. Thus, preparative plates can be used for separation of larger quantities of materials. The loading capacity for a given degree of sample component resolution increases roughly as the square root of the increased adsorbent layer thickness. In preparative TLC, materials to be separated are often applied as long streaks rather than spots. After development, specific compounds may be recovered by scraping the adsorbent layers from the plate in the region of interest and eluting the separated material from the adsorbent using a strong solvent. Thicker layers for higher capacity -Full range of plate sizes ·Prep-scored option for convenient zone location - Available with preadsorbent sample loading zone - Calcium sulfate binder-soft later for easy scraping