Learn More
Biotage TLC Plates, 2.5cm x 7.5 cm Glass, with silica, 100 per cas
SDP
Catalog No. 502312798
Encompass
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost.These glass plates are coated with silica and are 2.5cm x 7.5cm packed 100 plates to a case
Catalog No. 50-231-2798 Supplier Biotage Supplier No. TLC2575FI
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost.These glass plates are coated with silica and are 2.5cm x 7.5cm packed 100 plates to a case