Learn More
Biotage TLC Plates, 5cm x 10 cm Glass, with silica, 50 per cas
SDP
Catalog No. 502312796
Encompass
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost.These glass plates are coated with silica and are 5cm x 10cm packed 50 plates to a case.
Catalog No. 50-231-2796 Supplier Biotage Supplier No. TLC0510FI
Biotage TLC plates are the perfect tool to predict flash methods for Biotage Sfar columns. For synthetic chemists, the benefits of matched TLC plates and flash cartridges are better purification throughput, increased compound purity and yield, and reduced solvent cost.These glass plates are coated with silica and are 5cm x 10cm packed 50 plates to a case.