Learn More
Macherey-Nagel TLC POLYESTER SHEET 5X20 PK/50
Catalog No. NC2586331
Encompass_Preferred
TLC polyester sheets with polyamide 6 (PA6, ε-polycaprolactame). 5x20 cm, 0.1 mm thickness
Catalog No. NC2586331 Supplier Macherey-Nagel Supplier No. 803012
TLC polyester sheets with polyamide 6 (PA6, ε-polycaprolactame). 5x20 cm, 0.1 mm thickness