Learn More
MilliporeSigma™ Supelco™ TLC Saturation Pads
Catalog No. 111016153 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
11-101-6153 20 x 20 cm
11-101-1158 10 x 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6153 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2652251PAK
Add to Cart
Add to Cart
Pads improve reproducibility and reduce 'edge effects' when performing thin layer chromatography.
Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100/Pk.
For Use With (Application) TLC
Packing Material Pk. of 100 Ea.
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm