Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TLC-Silica gel 60 GF254, EMD-Millipore

Catalog No. M1116781000 Shop All MilliporeSigma Products
Change view
Click to view available options
1kg, Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
M1116781000 1kg, Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1116781000 Supplier MilliporeSigma™ Supplier No. 1.11678.1000

SiO2 - Loose sorbents for TLC plates

CAS: 7631-86-9
Solubility Information: Insoluble (20°C)
Melting Point: 1710°C
F.W.: 60.08
Bulk: Density: 200 to 800kg/m3
pH: 7 (100g/L, H2O, 20°C, slurry)

TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic Bottle
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.