Learn More
Camag Scientific Tlc Sprayer, Complete With Spray Head Type A And B, 100 Ml And 50 Ml Reagent Bottles
SDP
Catalog No. 501979319
Encompass_Preferred
The Tlc Sprayer With The Integrated Rechargeable Pump Unit Is Used For Manual Spraying Of Derivatization Reagents Onto Tlc Plates. Two Differents Spray Heads Are Available. For Solutions Of Regular Viscosity, E.G. Lower Alcohol Solutions, Spray Head A Is Suitable. For Solutions Of Higher Viscosity, E.G. Sulfuric Acid Reagents, The Spray Head B Is Recommended. Air Is Used As The Propellant.
Catalog No. 50-197-9319 Supplier Camag Scientific Supplier No. 022.6530
The Tlc Sprayer With The Integrated Rechargeable Pump Unit Is Used For Manual Spraying Of Derivatization Reagents Onto Tlc Plates. Two Differents Spray Heads Are Available. For Solutions Of Regular Viscosity, E.G. Lower Alcohol Solutions, Spray Head A Is Suitable. For Solutions Of Higher Viscosity, E.G. Sulfuric Acid Reagents, The Spray Head B Is Recommended. Air Is Used As The Propellant.