Learn More
Miles Scientific 10UL SYRINGE FOR SPOTON OR TLC AUTOSPOTTER 
SDP
Supplier:  Miles Scientific A8731
Encompass_Preferred
The TLC AUTOSPOTTER is a semi-automated device used to apply samples on Thin Layer Chromatography plates. The instrument eliminates the need for manual sample application and can be used to apply up to 18 samples at a time. The unit has been designed for use custom SGE brand TLC syringes with blunt Teflon tipped needles. These special needles minimize sample " creep back " and enhance reproducibility. The syringes also feature Teflon plunger tips to help eliminate the problem of metal-to-glass contamination. Syringes are available in 10, 25, 50, 100 and 250ul volumes. The device can apply samples at variable rates ranging from 3 minutes (fastest speed) to 30 minutes (slowest speed.)
Catalog No. NC9686871