Learn More
Enzo Life Sciences TLCK (200mg). CAS: 4238-41-9
SDP
Supplier:  Enzo Life Sciences BMLPI1210200
Encompass_Preferred
Irreversible inhibitor of trypsin and trypsin-like serine proteases such as granzyme D and tryptase. Also inhibits other proteases including plasmin, thrombin, and papain and some kinases including PKC. Inhibits LPS-induced expression of COX-2 in macrophages. Inhibits apoptosis (possibly through the inhibition of serine proteases). Chymotrypsin is not inhibited by TLCK, so this inhibitor can be used to block contaminating trypsin activity in chymoptrypsin preparations. TLCK can be used in combination with a chymotrypsin inhibitor to prevent proteolytic degradation during the isolation of proteins. Alternative name: N alpha-Tosyl-Lys-chloromethylketone . HCl. Purity: ≥95% (TLC). Solubility: Soluble in methanol (50mg/ml), DMSO (5mM) or water. Stock solutions at concentrations up to 10mM can be prepared in aqueous buffers with a pH of 6.0 or lower. TLCK is very unstable in buffers with a pH higher than 6 and will decompose rapidly. Long Term Storage: -20°C.
Catalog No. 50-201-1458