Learn More
Abcam TLCK (Tosyl-L-lysyl-chloromethane hydrochloride), Trypsin inhibitor, 200MG
Catalog No. NC1985022
Encompass_Preferred
MW 369.3 Da, Purity >95%. Irreversible inhibitor of trypsin and trypsin-like serine proteases. Blocks LPS- or cytokine-induced NF-κB activation. Inhibits caspase processing; blocks apoptosis. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1985022 Supplier Abcam Supplier No. AB144542200MG
MW 369.3 Da, Purity >95%. Irreversible inhibitor of trypsin and trypsin-like serine proteases. Blocks LPS- or cytokine-induced NF-κB activation. Inhibits caspase processing; blocks apoptosis. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement