Learn More
Aviva Systems Biology Corp TLE2 Antibody - N-terminal region (ARP32347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLE2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLE2. Alternate names: ESG, ESG2, FLJ41188, GRG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54538