Learn More
Aviva Systems Biology Corp TLE3 Antibody - N-terminal region (ARP32362_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32362_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TLE3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLE3. Alternate names: ESG, ESG3, GRG3, HsT18976ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54544