Learn More
Aviva Systems Biology Corp TLE4 Antibody - N-terminal region (ARP32381_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32381_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLE4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLE4. Alternate names: BCE-1, E(spI), ESG, ESG4, GRG4, BCE1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54555