Learn More
Carnabio Usa Inc TLK2/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 051675
Encompass
TLK2, Full-length, Wild type, Amino Acid 1-750, NP_006843.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0643