Learn More
Aviva Systems Biology Corp TLK2 Antibody - N-terminal region (ARP58343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TLK2. Alternate names: MGC44450, PKU-ALPHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-464