Learn More
Aviva Systems Biology Corp TLR2 Antibody - N-terminal region (ARP59037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TLR2. Alternate names: CD282, TIL4ApplicationIHC, WB, IF. Species reactivity: Dog, Human, Rat
Catalog No. 50-286-565