Learn More
Aviva Systems Biology Corp TLR2 Antibody - middle region (ARP59036_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59036_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TLR2. Alternate names: CD282, TIL4ApplicationIF, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-564