Learn More
Aviva Systems Biology Corp TLR2 Antibody - C-terminal region (ARP30551_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30551_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR2.The immunogen is a synthetic peptide directed towards the C terminal region of human TLR2. Alternate names: CD282, TIL4ApplicationIF, IHC-FFPE, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-014-54163