Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TLR2 (Human) ELISA Kit

Catalog No. 89100511 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological Fluid, CCS, Plasma, Serum, Tissue Homogenate
CCS, Plasma (EDTA, Heparin), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-511 CCS, Plasma (EDTA, Heparin), Serum
89-025-061 Biological Fluid, CCS, Plasma, Serum, Tissue Homogenate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-511 Supplier Abnova™ Supplier No. KA2350
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Immunoassay for quantitative detection of Human TLR2 in serum, plasma, cell culture supernatant

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description TLR2 (Human) ELISA Kit is an immunoassay for quantitative detection of human TLR2 in serum, plasma, cell culture supernatant.
Detection Limit <10 pg/mL
Detection Method Colorimetric
Includes Lyophilized recombinant human TLR2 standard (10ng/tube x 2 ), One 96-well plate precoated with anti-human TLR2 antibody, Sample diluent buffer (30mL), Biotinylatedanti-human TLR2 antibody, dilution 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100 (130μL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
Form Lyophilized
For Use With (Application) Quantification
Product Type TLR2 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.