Learn More
Invivogen LPS-B5 Standard, 5 mg
SDP
Catalog No. NC1008081
Encompass
LPS-B5 is a standard lipopolysaccharide (LPS) preparation extracted by a phenol-water mixture. LPS-B5 contains other bacterial components, such as lipoproteins, and therefore stimulates both TLR4 and TLR2.
Catalog No. NC1008081 Supplier Invivogen Supplier No. TLRLB5LPS
LPS-B5 is a standard lipopolysaccharide (LPS) preparation extracted by a phenol-water mixture. LPS-B5 contains other bacterial components, such as lipoproteins, and therefore stimulates both TLR4 and TLR2.