Learn More
Medchemexpress LLC TLR3 Human HEK293 10ug | 10ug
SDP
Catalog No. 5000187678
Encompass_Preferred
The TLR3 protein is a key Toll-like receptor family member that plays an important role in the innate immune system s recognition of pathogen-associated molecular patterns especially viral double-stranded RNA Its research enhances understanding of immune responses and provides potential applications for vaccine development and antiviral treatments TLR3 Protein Human (HEK293 His) is the recombinant human-derived TLR3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87678 Supplier Medchemexpress LLC Supplier No. HYP7785910UG
The TLR3 protein is a key Toll-like receptor family member that plays an important role in the innate immune system s recognition of pathogen-associated molecular patterns especially viral double-stranded RNA Its research enhances understanding of immune responses and provides potential applications for vaccine development and antiviral treatments TLR3 Protein Human (HEK293 His) is the recombinant human-derived TLR3 protein expressed by HEK293 with C-His labeled tag