Learn More
Enzo Life Sciences Toll-like receptor 3 monoclonal antibody (40C1285.6) (R-PE conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804362RC100
Encompass_Preferred
Clone: 40C1285.6. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 55-70 (VLNLTHNQLRRLPAAN) of human TLR3 (Toll-like receptor 3) cytoplasmic portion. Species reactivity: Human, Mouse, Dog. Applications: Flow Cytometry, ICC. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9885