Learn More
Medchemexpress LLC TLR4 Human sf9 Hi 10ug
SDP
Catalog No. 5000180740
Encompass_Preferred
TLR4 is a transmembrane receptor that recognizes PAMPs and DAMPs and cooperates with LY96 for LPS detection TLR4 activates MYD88-dependent and independent signaling pathways and participates in TIRAP TRAF6 and CD14-mediated endocytosis TLR4 Protein Human (sf9 His Solution) is the recombinant human-derived TLR4 protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-80740 Supplier Medchemexpress LLC Supplier No. HYP73586Y10UG
TLR4 is a transmembrane receptor that recognizes PAMPs and DAMPs and cooperates with LY96 for LPS detection TLR4 activates MYD88-dependent and independent signaling pathways and participates in TIRAP TRAF6 and CD14-mediated endocytosis TLR4 Protein Human (sf9 His Solution) is the recombinant human-derived TLR4 protein expressed by Sf9 insect cells with C-His labeled tag