Learn More
TARGETMOL CHEMICALS INC TLR4-IN-C34 25MG
SDP
Catalog No. 502576238
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TLR4-IN-C34 is an inhibitor of TLR4 and reduces systemic inflammation in models of endotoxemia and necrotizing enterocolitispurity: 99%
Catalog No. 50-257-6238 Supplier TARGETMOL CHEMICALS INC Supplier No. T850325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TLR4-IN-C34 is an inhibitor of TLR4 and reduces systemic inflammation in models of endotoxemia and necrotizing enterocolitispurity: 99%