Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tlr4-in-c34-c2-cooh | 1159408-54-4 | MFCD30802159 | 99.9% | 447.43 g/mol | C19H29NO11 | 100 MG
SDP

Catalog No. 5000209303
Encompass_Preferred

TLR4-IN-C34-C2-COOH is a small-molecule PROTAC linker that incorporates a TLR4 inhibitor moiety and is supplied for research use to support studies of TLR4 biology and probe or degrader synthesis. It has documented activity inhibiting TLR4 in enterocytes and macrophages and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis.

  • High purity of 99.9%.
  • Molecular formula C19H29NO11; molecular weight 447.43 g/mol.
  • Soluble in DMSO and water at 100 mg/mL with ultrasonic assistance.
  • Formulation examples yield ≥ 2.5 mg/mL (5.59 mM) in common in vivo vehicles.
  • Available in multiple pack sizes including 100 mg.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-09303 Supplier Medchemexpress LLC Supplier No. HYW092043100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR4-IN-C34-C2-COOH is a small-molecule PROTAC linker that incorporates a TLR4 inhibitor moiety and is supplied for research use to support studies of TLR4 biology and probe or degrader synthesis. It has documented activity inhibiting TLR4 in enterocytes and macrophages and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis.

  • High purity of 99.9%.
  • Molecular formula C19H29NO11; molecular weight 447.43 g/mol.
  • Soluble in DMSO and water at 100 mg/mL with ultrasonic assistance.
  • Formulation examples yield ≥ 2.5 mg/mL (5.59 mM) in common in vivo vehicles.
  • Available in multiple pack sizes including 100 mg.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.