Learn More
Medchemexpress LLC TLR4-in-c34-c2-cooh | 1159408-54-4 | MFCD30802159 | 99.9% | 447.43 g/mol | C19H29NO11 | 250 MG
SDP
Catalog No. 5000208858
Encompass_Preferred
TLR4-IN-C34-C2-COOH is a PROTAC linker that incorporates a TLR4 inhibitor and is supplied as a research-grade solid. It has defined physicochemical properties, including formula C19H29NO11, molecular weight 447.43 g/mol, high purity, and documented solubility profiles for in vitro and in vivo use.
  • Linker that incorporates a TLR4 inhibitor for modulation of toll-like receptor 4 signaling.
  • High purity (99.9%) and defined molecular weight and formula.
  • Solid form with solubility in DMSO and water at 100 mg/mL with ultrasonic agitation.
  • Available in multiple pack sizes, including 250 mg.
  • Intended for in vitro and in vivo research applications; for research use only.
Catalog No. 50-002-08858 Supplier Medchemexpress LLC Supplier No. 250MGHYW092043
TLR4-IN-C34-C2-COOH is a PROTAC linker that incorporates a TLR4 inhibitor and is supplied as a research-grade solid. It has defined physicochemical properties, including formula C19H29NO11, molecular weight 447.43 g/mol, high purity, and documented solubility profiles for in vitro and in vivo use.
  • Linker that incorporates a TLR4 inhibitor for modulation of toll-like receptor 4 signaling.
  • High purity (99.9%) and defined molecular weight and formula.
  • Solid form with solubility in DMSO and water at 100 mg/mL with ultrasonic agitation.
  • Available in multiple pack sizes, including 250 mg.
  • Intended for in vitro and in vivo research applications; for research use only.