Learn More
Aviva Systems Biology Corp TLR5 Antibody - N-terminal region (ARP31753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TLR5. Alternate names: FLJ10052, MGC126430, MGC126431, SLEB1, TIL3ApplicationWB. Species reactivity: Cow, Goat, Human, Pig, Rat, Sheep
Catalog No. 50-014-54320