Learn More
Aviva Systems Biology Corp TLR6 Antibody - middle region (ARP31755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TLR6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TLR6. Alternate names: CD286ApplicationIHC, WB. Species reactivity: Goat, Human, Sheep
Catalog No. 50-014-54321