Learn More
Enzo Life Sciences Toll-like receptor 6 (human) monoclonal antibody (86B1153.2) (FITC conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804363FC100
Encompass_Preferred
Clone: 86B1153.2. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 408-424 in the cytoplasmic portion of human TLR6 (Toll-like receptor 6). Species reactivity: Human. Applications: Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In 10mM TRIS containing 150mM sodium chloride and 0.05% sodium azide.
Catalog No. 50-200-9887