Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tlr7/8/9-in-1 | 2180127-82-4 | 98.5% | 435.60 g/mol | C27H37N3O2 | 5 MG
SDP

Catalog No. 5000262564
Encompass_Preferred

TLR7/8/9-IN-1 is a potent, orally bioavailable small-molecule antagonist of Toll-like receptors 7, 8, and 9 used in preclinical studies to inhibit TLR signaling and downstream cytokine release. The compound is supplied as a high-purity solid with published in vitro and in vivo solubility and activity data.

  • Potent antagonist activity (IC50 = 43 nM in TLR7/8/9 assays)
  • Demonstrates 4.8 nM potency versus TLR7 in PBMC IL-6 reduction
  • High reported purity of 98.5%
  • Molecular weight 435.60 g/mol; formula C27H37N3O2
  • Solid, off-white to light-yellow physical appearance
  • Soluble in DMSO (83.33 mg/mL) and formulated for in vivo use (≥2.08 mg/mL)
  • Suitable for preclinical studies of TLR pathway modulation

Catalog No. 50-002-62564 Supplier Medchemexpress LLC Supplier No. HY1319525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7/8/9-IN-1 is a potent, orally bioavailable small-molecule antagonist of Toll-like receptors 7, 8, and 9 used in preclinical studies to inhibit TLR signaling and downstream cytokine release. The compound is supplied as a high-purity solid with published in vitro and in vivo solubility and activity data.

  • Potent antagonist activity (IC50 = 43 nM in TLR7/8/9 assays)
  • Demonstrates 4.8 nM potency versus TLR7 in PBMC IL-6 reduction
  • High reported purity of 98.5%
  • Molecular weight 435.60 g/mol; formula C27H37N3O2
  • Solid, off-white to light-yellow physical appearance
  • Soluble in DMSO (83.33 mg/mL) and formulated for in vivo use (≥2.08 mg/mL)
  • Suitable for preclinical studies of TLR pathway modulation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.