Learn More
Medchemexpress LLC HY-103698A 5mg Medchemexpress, TLR7/8 agonist 1 (dihydrochloride) CAS:1620278-72-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103698A5MG
Encompass_Preferred
Medchemexpress, HY-103698A 5mg TLR7/8 agonist 1 (diHCl) CAS:1620278-72-9 TLR7/8 agonist 1 dihydrochloride is a toll-like receptor (TLR7)/TLR8 dual-agonistic imidazoquinoline. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8165