Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TLR7/8 agonist 1 dihydrochloride | 1620278-72-9 | MFCD31657341 | 99.7% | 432.39 | C22H27Cl2N5 | 10 MG
SDP

Catalog No. 5000243547
Encompass_Preferred

TLR7/8 agonist 1 dihydrochloride is a small-molecule imidazoquinoline research compound that acts as a dual agonist of Toll-like receptors 7 and 8. It is intended for use in immunology and inflammation studies and demonstrates low-nanomolar activity in receptor assays, suitable for both in vitro and in vivo experimental applications.

  • Dual agonist of TLR7 and TLR8 with low-nanomolar potency.
  • High reported purity (99.7%).
  • Molecular weight 432.39 and formula C22H27Cl2N5.
  • High solubility in DMSO (83.33 mg/mL) and compatible with common in vivo vehicles.
  • Supplied as an off-white to light yellow solid and available as a DMSO solution.
  • Storage: 4°C sealed; in solvent store at -80°C for long term.

Catalog No. 50-002-43547 Supplier Medchemexpress LLC Supplier No. HY103698A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7/8 agonist 1 dihydrochloride is a small-molecule imidazoquinoline research compound that acts as a dual agonist of Toll-like receptors 7 and 8. It is intended for use in immunology and inflammation studies and demonstrates low-nanomolar activity in receptor assays, suitable for both in vitro and in vivo experimental applications.

  • Dual agonist of TLR7 and TLR8 with low-nanomolar potency.
  • High reported purity (99.7%).
  • Molecular weight 432.39 and formula C22H27Cl2N5.
  • High solubility in DMSO (83.33 mg/mL) and compatible with common in vivo vehicles.
  • Supplied as an off-white to light yellow solid and available as a DMSO solution.
  • Storage: 4°C sealed; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.