Learn More
Medchemexpress LLC TLR7/8 agonist 1 dihydrochloride | 1620278-72-9 | MFCD31657341, MFCD26793840 | 99.7% | 432.39 | C22H27Cl2N5 | 50 MG
SDP
Catalog No. 5000244451
Encompass_Preferred
TLR7/8 agonist 1 dihydrochloride is an imidazoquinoline-based small molecule that functions as a dual agonist of Toll-like receptors 7 and 8. Supplied as a dihydrochloride salt in solid form, it is intended for research use in immunostimulation and TLR signaling studies and includes recommended solubility and storage guidance.
  • Dual TLR7/TLR8 agonist activity suitable for immunostimulation studies.
  • Dihydrochloride salt form improves solubility and handling.
  • High purity (99.7%) for reproducible experimental results.
  • Solid, off-white to light yellow appearance for easy visual inspection.
  • Highly soluble in DMSO; in vitro and in vivo formulation protocols provided.
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-44451 Supplier Medchemexpress LLC Supplier No. HY103698A50MG
TLR7/8 agonist 1 dihydrochloride is an imidazoquinoline-based small molecule that functions as a dual agonist of Toll-like receptors 7 and 8. Supplied as a dihydrochloride salt in solid form, it is intended for research use in immunostimulation and TLR signaling studies and includes recommended solubility and storage guidance.
  • Dual TLR7/TLR8 agonist activity suitable for immunostimulation studies.
  • Dihydrochloride salt form improves solubility and handling.
  • High purity (99.7%) for reproducible experimental results.
  • Solid, off-white to light yellow appearance for easy visual inspection.
  • Highly soluble in DMSO; in vitro and in vivo formulation protocols provided.
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).