Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TLR7/8 agonist 1 dihydrochloride | 1620278-72-9 | MFCD31657341; MFCD26793840 | 99.7% | 432.39 g/mol | C22H27Cl2N5 | 25 MG
SDP

Catalog No. 5000243266
Encompass_Preferred

TLR7/8 agonist 1 dihydrochloride is a TLR7/TLR8 dual-agonistic imidazoquinoline salt used as a research tool to activate innate immune signaling in cellular and animal studies. It is supplied as a high-purity solid suitable for in vitro and in vivo formulations.

  • High purity: 99.7%.
  • Molecular weight: 432.39 g/mol.
  • Chemical formula: C22H27Cl2N5.
  • Appearance: off-white to white solid.
  • Soluble in DMSO: 83.33 mg/mL (requires ultrasonic).
  • In vivo formulations: soluble ≥2.08 mg/mL in multiple vehicle systems.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: TLR7/8 activation in immunology and pharmacology studies.

Catalog No. 50-002-43266 Supplier Medchemexpress LLC Supplier No. HY103698A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7/8 agonist 1 dihydrochloride is a TLR7/TLR8 dual-agonistic imidazoquinoline salt used as a research tool to activate innate immune signaling in cellular and animal studies. It is supplied as a high-purity solid suitable for in vitro and in vivo formulations.

  • High purity: 99.7%.
  • Molecular weight: 432.39 g/mol.
  • Chemical formula: C22H27Cl2N5.
  • Appearance: off-white to white solid.
  • Soluble in DMSO: 83.33 mg/mL (requires ultrasonic).
  • In vivo formulations: soluble ≥2.08 mg/mL in multiple vehicle systems.
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: TLR7/8 activation in immunology and pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.