Learn More
Medchemexpress LLC TLR7/8 agonist 3 | 642473-95-8 | 100.0% | 50 MG
SDP
Catalog No. 5000338924
Encompass_Preferred
A potent agonist for TLR7 and TLR8, this compound is derived from patent WO2016057618 (compound of formula (II)) and is intended for research use only. It induces IFN-α and TNF-α in PBMC with EC50 values of 0.12 μM and 0.37 μM, respectively.
  • Potent agonist for TLR7 and TLR8.
  • Induces IFN-α and TNF-α.
  • Suitable for in vitro research applications.
Catalog No. 50-003-38924 Supplier Medchemexpress LLC Supplier No. HY13079750MG
A potent agonist for TLR7 and TLR8, this compound is derived from patent WO2016057618 (compound of formula (II)) and is intended for research use only. It induces IFN-α and TNF-α in PBMC with EC50 values of 0.12 μM and 0.37 μM, respectively.
  • Potent agonist for TLR7 and TLR8.
  • Induces IFN-α and TNF-α.
  • Suitable for in vitro research applications.