Learn More
Medchemexpress LLC TLR7/8 agonist 3 | 642473-95-8 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130797100MG
Encompass_Preferred
TLR7/8 agonist 3 (Compound II) is a potent TLR7 and TLR8 agonist, extracted from patent WO2016057618 (compound of formula (II)). It is intended for research use only.
  • Potent TLR7 and TLR8 agonist
  • Induces IFN-α and TNF-α in PBMC
  • For research use only
  • High purity (99.9%)
Catalog No. 50-003-39097