Learn More
Medchemexpress LLC Tlr7/8 agonist 4 | 2388520-33-8 | 99.2% | 324.42 | 250 MG
SDP
Catalog No. 5000351136
Encompass_Preferred
TLR7/8 agonist 4 (compound 41) is a potent Toll-like Receptor 7/8 agonist with anti-cancer activity. It is relevant for inducing potent anti-tumor immune responses by delivering neoantigens to activated dendritic cells via antibody-tumor immune complexes, helping to overcome the immune system's "immune ignorance" towards tumors. Toll-like receptor agonists, such as this product, are considered a key potential strategy in cancer treatment.
  • Potent TLR7/8 agonist
  • Exhibits anti-cancer activity
  • Induces potent anti-tumor immune responses
  • Helps overcome immune system's immune ignorance towards tumors
  • Considered a potential strategy in cancer treatment
Catalog No. 50-003-51136 Supplier Medchemexpress LLC Supplier No. 250MGHY139017
TLR7/8 agonist 4 (compound 41) is a potent Toll-like Receptor 7/8 agonist with anti-cancer activity. It is relevant for inducing potent anti-tumor immune responses by delivering neoantigens to activated dendritic cells via antibody-tumor immune complexes, helping to overcome the immune system's "immune ignorance" towards tumors. Toll-like receptor agonists, such as this product, are considered a key potential strategy in cancer treatment.
  • Potent TLR7/8 agonist
  • Exhibits anti-cancer activity
  • Induces potent anti-tumor immune responses
  • Helps overcome immune system's immune ignorance towards tumors
  • Considered a potential strategy in cancer treatment