Learn More
TARGETMOL CHEMICALS INC TLR7 8-IN-1 1MG
SDP
Catalog No. 502876237
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TLR7/8-IN-1, a crystalline TLR7/TLR8 inhibitor from patent WO2019220390 (compound 2b), is a valuable compound for autoimmune disease research. Purity 99.83%
Catalog No. 50-287-6237 Supplier TARGETMOL CHEMICALS INC Supplier No. T380781MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TLR7/8-IN-1, a crystalline TLR7/TLR8 inhibitor from patent WO2019220390 (compound 2b), is a valuable compound for autoimmune disease research. Purity 99.83%