Learn More
Medchemexpress LLC TLR7/8-IN-1 | 2205095-75-4 | 99.9% | 532.68 | 100 MG
SDP
Catalog No. 5000350503
Encompass_Preferred
TLR7/8-IN-1 is a crystalline form of a TLR7/TLR8 inhibitor. It is extracted from patent WO2019220390, compound 2b, and is intended for the research of autoimmune diseases. This product is for research use only.
  • Crystalline form
  • Functions as a TLR7/TLR8 inhibitor
  • Useful for autoimmune disease research
  • 99.9% purity
  • Molecular weight of 532.68
  • White to off-white solid appearance
Catalog No. 50-003-50503 Supplier Medchemexpress LLC Supplier No. HY139323100MG
TLR7/8-IN-1 is a crystalline form of a TLR7/TLR8 inhibitor. It is extracted from patent WO2019220390, compound 2b, and is intended for the research of autoimmune diseases. This product is for research use only.
  • Crystalline form
  • Functions as a TLR7/TLR8 inhibitor
  • Useful for autoimmune disease research
  • 99.9% purity
  • Molecular weight of 532.68
  • White to off-white solid appearance